Learn More
Waters Corp Waters ACQUITY Premier Peptide BEH C18 Column, 300Å, 1.7 μm, 2.1 x 150 mm
Supplier:  Waters Corp 186009647
Encompass_Preferred
This product is a method validation kit containing three columns, specifically designed for peptide separations. It utilizes wide-pore (300Å) trifunctionally bonded BEH particles, offering a broad usable pH range, excellent low pH stability, and minimal column bleed. This makes it suitable for analyzing large peptides in proteomics, protein characterization, and peptide synthesis, particularly with low dispersion systems for LC-MS methods.
  • Optimized and quality control tested with standards for peptide separations.
  • Offers a wide usable pH range and superior low pH stability.
  • Provides ultra-low column bleed for analyzing large peptides.
  • Enhances reproducibility, peak shape, and recovery for metal-sensitive analytes.
  • Recommended for use with low dispersion systems.
Catalog No. 50-203-9131
Add to Cart
Request a Quote