Learn More
Waters Corp Waters Corporation C18 Column, 1.7 μm, 2.1 x 50 mm
Catalog No. 502039087 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These Peptide Separation Technology Columns are optimized and QC tested for peptide separations. With a 130Å small-pore BEH particle, they offer a wide pH range, superior low pH stability, and ultra-low column bleed. Suitable for UPLC, UHPLC, and LC-MS methods, these columns enhance reproducibility, peak shape, and recovery for metal-sensitive analytes.
  • Optimized and QC tested for peptide separations.
  • Features a 130Å small-pore BEH particle, offering wide pH range, superior low pH stability, and ultra-low column bleed.
  • Utilizes MaxPeak High Performance Surfaces (HPS) technology, enhancing reproducibility and peak shape for metal-sensitive analytes.
  • Reduces column conditioning and passivation times, enabling simpler mobile phases with improved sensitivity.
  • Designed for peptide analysis, providing outstanding peak capacity superior peak shape.
Catalog No. 50-203-9087 Supplier Waters Corp Supplier No. 186009481
These Peptide Separation Technology Columns are optimized and QC tested for peptide separations. With a 130Å small-pore BEH particle, they offer a wide pH range, superior low pH stability, and ultra-low column bleed. Suitable for UPLC, UHPLC, and LC-MS methods, these columns enhance reproducibility, peak shape, and recovery for metal-sensitive analytes.
  • Optimized and QC tested for peptide separations.
  • Features a 130Å small-pore BEH particle, offering wide pH range, superior low pH stability, and ultra-low column bleed.
  • Utilizes MaxPeak High Performance Surfaces (HPS) technology, enhancing reproducibility and peak shape for metal-sensitive analytes.
  • Reduces column conditioning and passivation times, enabling simpler mobile phases with improved sensitivity.
  • Designed for peptide analysis, providing outstanding peak capacity superior peak shape.