Learn More
Waters Corp Column, CSH C18, 1.7 μm, 2.1 x 100 mm, 130 Å
Supplier:  Waters Corp 186010710
Encompass_Preferred
  • Engineered for peptide separations.
  • Provides excellent peak shape with FA- or TFA-containing mobile phases.
  • Suitable for LC/LC-MS applications; low dependence on strong ion pairing agents.
  • QC tested with cytochrome c digest standards.
  • 2.1 mm ID recommended for low dispersion UPLC/UHPLC and LC-MS methods.
  • Enhances reproducibility, peak shape, and recovery for metal-sensitive analytes.
  • Features integrated, replaceable 5 mm pre-column.
  • Pre-column extends column life without impacting resolution.
  • Replacements use same qualified sorbent.
Catalog No. 50-239-1676