Learn More
Waters Corp Column, CSH C18, 130 Å, 1.7 μm, 2.1 x 150 mm
Supplier:  Waters Corp 186010711
Encompass_Preferred
This column is designed for peptide separations, incorporating particles with low, carefully-defined positive charges. This ensures excellent peak shape for peptides with FA- or TFA-containing mobile phases, and is quality controlled using cytochrome c digest standards. It is suitable for LC or LC-MS applications, especially with low dispersion (UPLC or UHPLC) systems for LC-MS methods.
  • Ensures excellent peak shape for peptides and minimal reliance on strong ion pairing agents.
  • Improves reproducibility, peak shape, and recovery for metal-sensitive analytes.
  • Features an integrated, easily replaceable 5 mm pre-column to protect and extend analytical column life.
  • Replacement cartridges contain the same qualified sorbent.
Catalog No. 50-239-2503