Learn More
Waters Corp Column, Peptide HSS T3, 100 Å, 1.8 μm, 2.1 × 100 mm
Catalog No. 502391410 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY Premier Peptide HSS T3 column is engineered for the separation of small, polar peptides, providing superior retentivity compared to traditional hybrid particle technologies. It integrates MaxPeak High Performance Surfaces (HPS) to reduce problematic analyte/surface interactions, ensuring enhanced recovery, sensitivity, and reproducibility in peptide analysis.
  • Minimizes analyte/surface interactions to prevent sample loss.
  • Increases analyte recovery, sensitivity, and reproducibility.
  • Mitigates the loss of metal-sensitive analytes.
  • Reduces column conditioning and passivation times.
  • Improves sensitivity and peak shapes.
  • Allows for the use of simpler mobile phases.
  • Saves time in method development.
  • Enhances data confidence and reduces risk.
Catalog No. 50-239-1410 Supplier Waters Corp Supplier No. 186010722
The ACQUITY Premier Peptide HSS T3 column is engineered for the separation of small, polar peptides, providing superior retentivity compared to traditional hybrid particle technologies. It integrates MaxPeak High Performance Surfaces (HPS) to reduce problematic analyte/surface interactions, ensuring enhanced recovery, sensitivity, and reproducibility in peptide analysis.
  • Minimizes analyte/surface interactions to prevent sample loss.
  • Increases analyte recovery, sensitivity, and reproducibility.
  • Mitigates the loss of metal-sensitive analytes.
  • Reduces column conditioning and passivation times.
  • Improves sensitivity and peak shapes.
  • Allows for the use of simpler mobile phases.
  • Saves time in method development.
  • Enhances data confidence and reduces risk.