Learn More
Waters Corp ACQUITY Premier Peptide HSS T3 Column, 100 Å, 1.8 μm, 2.1 x 150 mm
Catalog No. 502392727 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column is designed for separations requiring silica-based selectivities or increased retention of hydrophilic peptides. It delivers chromatographic performance from particle technologies, enhancing reproducibility, peak shape, and recovery for metal-sensitive analytes.
  • Each batch of HSS T3, 100Å material is QC tested with a gradient separation of a tryptic digest of cytochrome c and must pass stringent performance criteria.
  • 2.1 mm ID columns are recommended for use with low dispersion (UPLC or UHPLC) systems, especially in LC-MS methods.
  • Unique column design includes a fully integrated and easily replaceable 5 mm pre-column that protects and extends the life of the analytical column without adding dispersion or impacting resolution.
  • Ideal for separation of small, polar peptides.
  • Offers greater retentivity compared to hybrid particle technology columns.
Catalog No. 50-239-2727 Supplier Waters Corp Supplier No. 186010723
This column is designed for separations requiring silica-based selectivities or increased retention of hydrophilic peptides. It delivers chromatographic performance from particle technologies, enhancing reproducibility, peak shape, and recovery for metal-sensitive analytes.
  • Each batch of HSS T3, 100Å material is QC tested with a gradient separation of a tryptic digest of cytochrome c and must pass stringent performance criteria.
  • 2.1 mm ID columns are recommended for use with low dispersion (UPLC or UHPLC) systems, especially in LC-MS methods.
  • Unique column design includes a fully integrated and easily replaceable 5 mm pre-column that protects and extends the life of the analytical column without adding dispersion or impacting resolution.
  • Ideal for separation of small, polar peptides.
  • Offers greater retentivity compared to hybrid particle technology columns.