Learn More
Waters Corp ACQUITY Premier Peptide HSS T3 Column, 100Å, 1.8 μm, 2.1 x 50 mm
Catalog No. 502039091 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY Premier Peptide HSS T3, 100Å, 1.8 μm, 2.1 x 50 mm Column is designed for separations requiring silica-based selectivities or enhanced retention of hydrophilic peptides. It is QC tested for stringent performance and ideal for low dispersion UPLC or UHPLC systems, especially for LC-MS methods. Part of the MaxPeak Premier class, it improves reproducibility, peak shape, and recovery for metal-sensitive analytes.
  • Designed for increased retention of hydrophilic peptides.
  • Rigorously QC tested for stringent performance.
  • Ideal for low dispersion UPLC/UHPLC systems and LC-MS.
  • Enhances reproducibility, peak shape, and recovery for metal-sensitive analytes.
  • Employs High-Strength Silica (HSS) particle technology.
  • Compatible with 100% aqueous mobile phases.
  • Provides superior peak shape and batch-to-batch data reproducibility.
Catalog No. 50-203-9091 Supplier Waters Corp Supplier No. 186009490
The ACQUITY Premier Peptide HSS T3, 100Å, 1.8 μm, 2.1 x 50 mm Column is designed for separations requiring silica-based selectivities or enhanced retention of hydrophilic peptides. It is QC tested for stringent performance and ideal for low dispersion UPLC or UHPLC systems, especially for LC-MS methods. Part of the MaxPeak Premier class, it improves reproducibility, peak shape, and recovery for metal-sensitive analytes.
  • Designed for increased retention of hydrophilic peptides.
  • Rigorously QC tested for stringent performance.
  • Ideal for low dispersion UPLC/UHPLC systems and LC-MS.
  • Enhances reproducibility, peak shape, and recovery for metal-sensitive analytes.
  • Employs High-Strength Silica (HSS) particle technology.
  • Compatible with 100% aqueous mobile phases.
  • Provides superior peak shape and batch-to-batch data reproducibility.