Learn More
Waters Corp ACQUITY Premier Peptide HSS T3, 100 Å, 1.8 μm, 2.1 x 150 mm
Catalog No. 502039140 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This ACQUITY Premier Peptide HSS T3 column uses High Strength Silica particle technology for enhanced peptide retention and unique selectivity. It incorporates MaxPeak High Performance Surfaces for improved hardware inertness, reducing the number of injections required for desired performance.
  • Utilizes high strength silica particle technology for increased peptide retention and unique selectivity.
  • Manufactured in a cGMP, ISO 9001:2000 certified plant with stringent QC testing.
  • Designed and tested for maximum chromatographic performance on UPLC systems.
  • Incorporates MaxPeak™ high performance surfaces for improved hardware inertness.
  • Features eCord™ intelligent chip technology for comprehensive column performance data.
  • Includes a certificate of analysis and performance test chromatogram.
Catalog No. 50-203-9140 Supplier Waters Corp Supplier No. 186009656
Add to Cart
Add to Cart
Request a Quote
This ACQUITY Premier Peptide HSS T3 column uses High Strength Silica particle technology for enhanced peptide retention and unique selectivity. It incorporates MaxPeak High Performance Surfaces for improved hardware inertness, reducing the number of injections required for desired performance.
  • Utilizes high strength silica particle technology for increased peptide retention and unique selectivity.
  • Manufactured in a cGMP, ISO 9001:2000 certified plant with stringent QC testing.
  • Designed and tested for maximum chromatographic performance on UPLC systems.
  • Incorporates MaxPeak™ high performance surfaces for improved hardware inertness.
  • Features eCord™ intelligent chip technology for comprehensive column performance data.
  • Includes a certificate of analysis and performance test chromatogram.