Learn More
Waters Corp Waters Column, ACQUITY Premier Peptide HSS T3, 100Å Pore, 1.8 μm Particle, 2.1 x 150 mm
Catalog No. 502039093 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These columns are designed for peptide separations, offering silica-based selectivities and enhanced retention for hydrophilic peptides. They are part of the MaxPeak Premier class, improving reproducibility, peak shape, and recovery for metal-sensitive analytes in UPLC/UHPLC and LC-MS applications.
  • Designed for silica-based selectivities or increased retention of hydrophilic peptides.
  • HSS T3, 100Å material is QC tested with cytochrome c tryptic digest for stringent performance.
  • 2.1 mm ID columns are recommended for UPLC/UHPLC and LC-MS methods.
  • MaxPeak High Performance Surfaces maximize analyte recovery, sensitivity, and repeatability.
  • 100% water wettable, compatible with 100% aqueous mobile phases.
Catalog No. 50-203-9093 Supplier Waters Corp Supplier No. 186009492
These columns are designed for peptide separations, offering silica-based selectivities and enhanced retention for hydrophilic peptides. They are part of the MaxPeak Premier class, improving reproducibility, peak shape, and recovery for metal-sensitive analytes in UPLC/UHPLC and LC-MS applications.
  • Designed for silica-based selectivities or increased retention of hydrophilic peptides.
  • HSS T3, 100Å material is QC tested with cytochrome c tryptic digest for stringent performance.
  • 2.1 mm ID columns are recommended for UPLC/UHPLC and LC-MS methods.
  • MaxPeak High Performance Surfaces maximize analyte recovery, sensitivity, and repeatability.
  • 100% water wettable, compatible with 100% aqueous mobile phases.