Learn More
Waters Corp Column, BEH C4, 1.7 μm, 300 Å, 2.1 x 100 mm
Supplier:  Waters Corp 186010327
Encompass_Preferred
The ACQUITY Premier Protein BEH C4 column is optimized for separating proteins by size, hydrophobicity, and isoelectric point. It's a MaxPeak Premier Class column, enhancing reproducibility, peak shape, and recovery for metal-sensitive analytes. This chemistry is beneficial for low-level intact protein, subunit, and phosphorylated protein analysis, offering improved MS sensitivity.
  • Efficiently separates proteins by size, hydrophobicity, and isoelectric point.
  • C4 ligand minimizes protein carryover by being less retentive than C18 phases.
  • Enhances protein recovery and increases peak capacity.
  • Optimized for MS sensitivity at low levels.
  • Delivers expected chromatographic performance, improving reproducibility, peak shape, and recovery for metal-sensitive analytes.
  • 2.1mm ID columns recommended for low dispersion UPLC or UHPLC systems, especially for LC-MS methods.
Catalog No. 50-239-0695