Learn More
Waters Corp ACQUITY Premier Protein BEH C4, 300 Å, 1.7 μm, 2.1 x 50 mm
Catalog No. 502392308 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This chromatography column is an ACQUITY Premier Protein BEH C4 column. It is designed for analytical applications requiring specific pore and particle sizes to ensure high-performance separations.
  • Pore size of 300 Å.
  • Particle size of 1.7 μm.
  • Column dimensions of 2.1 x 50 mm.
  • Supplied as 1 per pack.
  • Available with a MassPREP Protein Mix Standard.
Catalog No. 50-239-2308 Supplier Waters Corp Supplier No. 176005107
This chromatography column is an ACQUITY Premier Protein BEH C4 column. It is designed for analytical applications requiring specific pore and particle sizes to ensure high-performance separations.
  • Pore size of 300 Å.
  • Particle size of 1.7 μm.
  • Column dimensions of 2.1 x 50 mm.
  • Supplied as 1 per pack.
  • Available with a MassPREP Protein Mix Standard.