Learn More
Waters Corp Column, Protein BEH C4, 300 Å, 1.7 μm, 2.1 x 100 mm
Supplier:  Waters Corp 176005108
Encompass_Preferred
ACQUITY Premier Protein BEH C4 300 Å columns are QC tested with a protein mix standard. They are optimized for separating proteins by size, hydrophobicity, and isoelectric point. The C4 ligand minimizes protein carryover, increases protein recovery, and improves peak capacity for metal-sensitive analytes.
  • Optimized for separating proteins based on size, hydrophobicity, and isoelectric point.
  • C4 ligand minimizes protein carryover, increases protein recovery, and improves peak capacity.
  • Enhanced reproducibility, peak shape, and recovery for metal-sensitive analytes.
  • Quality control tested with a protein mix standard.
  • Tolerates extreme pH and temperature conditions.
  • Provides minimal secondary interactions for protein analysis.
  • Enhances sensitivity for phosphorylated proteins and low-level intact and subunit monoclonal antibody analyses.
Catalog No. 50-239-2906