Learn More
Waters Corp ACQUITY Premier Protein SEC Column, 250 Å, 1.7 μm, 4.6 x 300 mm
Catalog No. 502390672 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY Premier Protein SEC 250Å, 1.7 μm Columns are designed for reliable protein size variant separations. They combine MaxPeak Premier High Performance Surfaces and BEH-PEO Particle technologies to minimize undesirable protein-column interactions, reducing method development needs for a wide range of samples.
  • Separates proteins ranging from 10,000 to 650,000 Daltons.
  • Reduces costly method development by minimizing secondary interactions.
  • Includes a guard column for extended column life.
  • Engineered for UPLC SEC applications with low LC system volume.
  • Operates within a pH range of 2.5 to 8 pH and up to 60 °C.
  • Maximum pressure rating of 11000 PSI (759 bar).
Catalog No. 50-239-0672 Supplier Waters Corp Supplier No. 176004784
Add to Cart
Add to Cart
Request a Quote
ACQUITY Premier Protein SEC 250Å, 1.7 μm Columns are designed for reliable protein size variant separations. They combine MaxPeak Premier High Performance Surfaces and BEH-PEO Particle technologies to minimize undesirable protein-column interactions, reducing method development needs for a wide range of samples.
  • Separates proteins ranging from 10,000 to 650,000 Daltons.
  • Reduces costly method development by minimizing secondary interactions.
  • Includes a guard column for extended column life.
  • Engineered for UPLC SEC applications with low LC system volume.
  • Operates within a pH range of 2.5 to 8 pH and up to 60 °C.
  • Maximum pressure rating of 11000 PSI (759 bar).