Learn More
Waters Corp ACQUITY Premier Protein SEC Column, 250Å, 1.7 μm, 4.6 x 150 mm
Supplier: Waters Corp 176004794
ACQUITY Premier Protein SEC 250Å, 1.7 μm Columns are engineered for reliable protein size variant separations across a molecular weight range of 10,000 to 650,000 Daltons. These columns utilize MaxPeak Premier High Performance Surfaces and BEH-PEO Particle technologies to minimize secondary ionic or hydrophobic interactions, simplifying method development and allowing for a generic method approach.
- Enables protein size variant separations up to 650,000 Daltons.
- MaxPeak Premier High Performance Surfaces and BEH-PEO Particle technology.
- Minimizes secondary interactions, simplifying method development.
- Includes mAb Size Variant Standard and a MaxPeak Premier SEC Guard.
- Designed for UPLC SEC applications.
- Pore size: 250 Å; Particle size: 1.7 μm.
- Dimensions: 4.6 mm ID x 150 mm L.
- pH range: 2.5 to 8; Max pressure: 7000 psi.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.