Learn More
Waters Corp Waters ACQUITY Premier Oligonucleotide C18 Column, 1.7 μm, 130 Å, 2.1 × 100 mm
Catalog No. NC1842937 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY Premier Oligonucleotide columns are designed for the separation of detritylated synthetic oligonucleotides using ion-pair, reversed-phase chromatography. QC testing with Waters MassPrep OST standards ensures consistent performance. As part of the MaxPeak Premier Class, these columns enhance reproducibility, peak shape, and recovery for metal-sensitive analytes.
  • C18 chemistry, reversed-phase separation
  • Hybrid, spherical particle substrate: 1.7 μm particle size, 130 Å pore size
  • pH range of 1 to 12; Max pressure 18000 psi (1240 Bar)
  • Endcapped, low silanol activity, Parker-style endfitting
  • QC tested for demanding oligonucleotide assays
  • Optimized for UPLC systems with BEH particle technology
  • Dimensions: 2.1 mm ID, 100 mm length; 18% carbon load, eCord enabled
  • Ideal for oligonucleotide and gene therapeutics
Catalog No. NC1842937 Supplier Waters Corp Supplier No. 186009485
ACQUITY Premier Oligonucleotide columns are designed for the separation of detritylated synthetic oligonucleotides using ion-pair, reversed-phase chromatography. QC testing with Waters MassPrep OST standards ensures consistent performance. As part of the MaxPeak Premier Class, these columns enhance reproducibility, peak shape, and recovery for metal-sensitive analytes.
  • C18 chemistry, reversed-phase separation
  • Hybrid, spherical particle substrate: 1.7 μm particle size, 130 Å pore size
  • pH range of 1 to 12; Max pressure 18000 psi (1240 Bar)
  • Endcapped, low silanol activity, Parker-style endfitting
  • QC tested for demanding oligonucleotide assays
  • Optimized for UPLC systems with BEH particle technology
  • Dimensions: 2.1 mm ID, 100 mm length; 18% carbon load, eCord enabled
  • Ideal for oligonucleotide and gene therapeutics