Learn More
Waters Corp Waters Column, ACQUITY Premier Peptide BEH C18, 130Å 1.7 μm, 2.1 x 100 mm
Catalog No. NC1842932 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These ACQUITY Premier Peptide BEH C18 Columns are optimized for peptide separations, offering a wide pH range and superior low pH stability. They are ideal for UPLC or UHPLC systems, enhancing reproducibility and peak shape for metal-sensitive analytes.
  • MaxPeak High Performance Surface hardware improves peak shape and signal intensity by reducing analyte/surface interactions.
  • Wide pH range (pH 1–12) and superior low pH stability.
  • C18 stationary phase with 1.7 μm particle size for high selectivity and sensitivity in peptide characterization.
  • 130Å pore size enables optimal sample penetration.
  • Offers exceptional performance, reproducibility, and long column lifetime.
  • Withstands high-pressure and is compatible with diverse mobile phases.
Catalog No. NC1842932 Supplier Waters Corp Supplier No. 186009482
These ACQUITY Premier Peptide BEH C18 Columns are optimized for peptide separations, offering a wide pH range and superior low pH stability. They are ideal for UPLC or UHPLC systems, enhancing reproducibility and peak shape for metal-sensitive analytes.
  • MaxPeak High Performance Surface hardware improves peak shape and signal intensity by reducing analyte/surface interactions.
  • Wide pH range (pH 1–12) and superior low pH stability.
  • C18 stationary phase with 1.7 μm particle size for high selectivity and sensitivity in peptide characterization.
  • 130Å pore size enables optimal sample penetration.
  • Offers exceptional performance, reproducibility, and long column lifetime.
  • Withstands high-pressure and is compatible with diverse mobile phases.