Learn More
Waters Corp Waters Corporation ACQUITY Premier Protein BEH C4 Column, 300 Å, 1.7 μm, 2.1 mm x 150 mm
Catalog No. NC2167692 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters Corporation ACQUITY Premier Protein BEH C4 Column is designed for reversed-phase chromatography (RPLC) applications, particularly for the analysis of AAV capsid proteins. It incorporates BEH particle technology and MaxPeak High Performance Surfaces to improve analyte recoveries and peak shapes.
  • Utilizes BEH (Ethylene Bridged Hybrid) particle technology.
  • Incorporated into Premier column hardware with MaxPeak High Performance Surfaces.
  • MaxPeak High Performance Surfaces provide higher analyte recoveries and improved peak shapes.
  • Pore size: 300 Å.
  • Particle size: 1.7 μm.
  • Dimensions: 2.1 mm diameter x 150 mm length.
  • Recommended for AAV capsid protein analysis using RPLC.
  • Used with DFA ion pairing.
  • Specifically shown for RPLC of AAV8 capsid proteins with DFA modified mobile phases.
This item is not returnable. View return policy
Catalog No. NC2167692 Supplier Waters Corp Supplier No. 186010328
The Waters Corporation ACQUITY Premier Protein BEH C4 Column is designed for reversed-phase chromatography (RPLC) applications, particularly for the analysis of AAV capsid proteins. It incorporates BEH particle technology and MaxPeak High Performance Surfaces to improve analyte recoveries and peak shapes.
  • Utilizes BEH (Ethylene Bridged Hybrid) particle technology.
  • Incorporated into Premier column hardware with MaxPeak High Performance Surfaces.
  • MaxPeak High Performance Surfaces provide higher analyte recoveries and improved peak shapes.
  • Pore size: 300 Å.
  • Particle size: 1.7 μm.
  • Dimensions: 2.1 mm diameter x 150 mm length.
  • Recommended for AAV capsid protein analysis using RPLC.
  • Used with DFA ion pairing.
  • Specifically shown for RPLC of AAV8 capsid proteins with DFA modified mobile phases.