Learn More
Waters Corp ACQUITY QDa II Mass Detector, Single Quadrupole, m/z 30 to 1250
Catalog No. NC3397552 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY QDa II Mass Detector is a single quadrupole mass detector that enhances chromatography systems by providing mass detection, minimizing the risk of unexpected components or co-elutions, and confirming trace sample constituents. It is designed to be easy to use, robust, compact, stackable, and energy-efficient. It aims to increase productivity and provide additional information from separations without the complexity or cost of traditional mass spectrometry.
  • Enhanced sensitivity, robustness, and wider mass range.
  • Integrates with UPLC, Arc, and HPLC systems.
  • Supports positive and negative electrospray ionization.
  • Typical mass range of m/z 30 to 1250.
  • Low maintenance and user-friendly design.
  • ACT Environmental Impact Factor Label for energy efficiency.
  • Automated mass calibration and resolution verification.
Catalog No. NC3397552 Supplier Waters Corp Supplier No. 176005407
The ACQUITY QDa II Mass Detector is a single quadrupole mass detector that enhances chromatography systems by providing mass detection, minimizing the risk of unexpected components or co-elutions, and confirming trace sample constituents. It is designed to be easy to use, robust, compact, stackable, and energy-efficient. It aims to increase productivity and provide additional information from separations without the complexity or cost of traditional mass spectrometry.
  • Enhanced sensitivity, robustness, and wider mass range.
  • Integrates with UPLC, Arc, and HPLC systems.
  • Supports positive and negative electrospray ionization.
  • Typical mass range of m/z 30 to 1250.
  • Low maintenance and user-friendly design.
  • ACT Environmental Impact Factor Label for energy efficiency.
  • Automated mass calibration and resolution verification.