Learn More
Waters Corp Maintenance Kit, ACQUITY QDa Performance, KAD, Annual
Catalog No. 502392971 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Performance Maintenance Kit is designed for ACQUITY QDa Standard (KAD serial numbers) instruments. It contains all the necessary quality parts to ensure the instrument operates at peak performance on an annual basis.
  • Kit includes seals and gaskets, calibration pin, aperture disk assembly, air filters, and rotary pump maintenance materials.
  • Ensures peak instrument performance when replaced annually.
  • Supports various system types including HPLC, MS, and UPLC.
  • Compatible with Arc Premier System, QDa Mass Detector, UPLC H-Class System, UPLC I-Class System, and Alliance HPLC System instruments.
  • One unit per package.
Catalog No. 50-239-2971 Supplier Waters Corp Supplier No. 201000309
This Performance Maintenance Kit is designed for ACQUITY QDa Standard (KAD serial numbers) instruments. It contains all the necessary quality parts to ensure the instrument operates at peak performance on an annual basis.
  • Kit includes seals and gaskets, calibration pin, aperture disk assembly, air filters, and rotary pump maintenance materials.
  • Ensures peak instrument performance when replaced annually.
  • Supports various system types including HPLC, MS, and UPLC.
  • Compatible with Arc Premier System, QDa Mass Detector, UPLC H-Class System, UPLC I-Class System, and Alliance HPLC System instruments.
  • One unit per package.