Learn More
Waters Corp Accumuator Head Rebuild Kit, HPLC System Type, Annual Replacement
Catalog No. 502392613 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This rebuild kit contains the plunger, seals, and check valve necessary to rebuild an ACQUITY H-Class Quaternary Solvent Manager Accumulator Pumphead.
  • Contains plunger, seals, and check valve.
  • Designed for rebuilding an ACQUITY H-Class Quaternary Solvent Manager Accumulator Pumphead.
  • System type: HPLC.
  • Replacement schedule: Annual.
Catalog No. 50-239-2613 Supplier Waters Corp Supplier No. 700011073
This rebuild kit contains the plunger, seals, and check valve necessary to rebuild an ACQUITY H-Class Quaternary Solvent Manager Accumulator Pumphead.
  • Contains plunger, seals, and check valve.
  • Designed for rebuilding an ACQUITY H-Class Quaternary Solvent Manager Accumulator Pumphead.
  • System type: HPLC.
  • Replacement schedule: Annual.