Learn More
Waters Corp Waters LockMass Kit, LC/MS, 12 Months Shelf Life, Room Temperature
Catalog No. NC1863660 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY RDa Waters_connect LockMass Kit is designed to compensate for mass shifts that occur due to changing lab temperatures, particularly for the BioAccord system. It contains a LockMass Concentrate and a Diluent. This kit ensures precise mass measurements by calibrating the mass spectrometer, regardless of temperature variations.
  • Compensates for mass shifts caused by changing lab temperatures.
  • Simple preparation by adding LockMass Concentrate to Diluent and mixing.
  • Stable for 12 months when stored at room temperature.
  • Ensures precise measurements and reproducible data.
  • Contains LeuEnk, Caffeine, and Pentenasulfonic Acid for effective calibration.
  • Diluent is a solution of water and methanol.
Catalog No. NC1863660 Supplier Waters Corp Supplier No. 186009298
The ACQUITY RDa Waters_connect LockMass Kit is designed to compensate for mass shifts that occur due to changing lab temperatures, particularly for the BioAccord system. It contains a LockMass Concentrate and a Diluent. This kit ensures precise mass measurements by calibrating the mass spectrometer, regardless of temperature variations.
  • Compensates for mass shifts caused by changing lab temperatures.
  • Simple preparation by adding LockMass Concentrate to Diluent and mixing.
  • Stable for 12 months when stored at room temperature.
  • Ensures precise measurements and reproducible data.
  • Contains LeuEnk, Caffeine, and Pentenasulfonic Acid for effective calibration.
  • Diluent is a solution of water and methanol.