Learn More
Waters Corp Waters Corporation Performance Maintenance Kit, Annual Replacement, UPLC System
Catalog No. 50466523 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Performance Maintenance Kit for the ACQUITY UPLC Sample Organizer includes all the necessary quality parts to ensure the instrument operates at peak performance on an annual basis.
  • Ensures instrument operates at peak performance.
  • Designed for UPLC systems.
  • Compatible with ACQUITY Arc Premier System, ACQUITY UPLC H-Class System, and ACQUITY instruments.
  • Recommended for annual replacement.
  • Includes air filters.
Catalog No. 50-466-523 Supplier Waters Corp Supplier No. 201000208
The Performance Maintenance Kit for the ACQUITY UPLC Sample Organizer includes all the necessary quality parts to ensure the instrument operates at peak performance on an annual basis.
  • Ensures instrument operates at peak performance.
  • Designed for UPLC systems.
  • Compatible with ACQUITY Arc Premier System, ACQUITY UPLC H-Class System, and ACQUITY instruments.
  • Recommended for annual replacement.
  • Includes air filters.