Learn More
Waters Corp Performance Maintenance Kit, Annual, ACQUITY Binary Solvent Manager, UPLC System
Catalog No. 502392721 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Performance Maintenance Kit for the ACQUITY Binary Solvent Manager contains all the Quality Parts™ necessary to keep your instrument operating at peak performance on an annual basis.
  • Contains solvent filters
  • Includes in-line filters
  • Features tubing
  • Includes air filter chassis
  • Contains check valves
  • Features plungers
  • Includes plunger seals
  • Contains seal wash seals
  • Designed for UPLC system applications
  • Compatible with ACQUITY Arc Premier systems
  • Recommended for annual replacement
Catalog No. 50-239-2721 Supplier Waters Corp Supplier No. 201000173
This Performance Maintenance Kit for the ACQUITY Binary Solvent Manager contains all the Quality Parts™ necessary to keep your instrument operating at peak performance on an annual basis.
  • Contains solvent filters
  • Includes in-line filters
  • Features tubing
  • Includes air filter chassis
  • Contains check valves
  • Features plungers
  • Includes plunger seals
  • Contains seal wash seals
  • Designed for UPLC system applications
  • Compatible with ACQUITY Arc Premier systems
  • Recommended for annual replacement