Learn More
Waters Corp Waters Flowcell, ACQUITY TUV, 10 mm, 500 nL
Catalog No. 50179743 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This TUV flow cell is a low-dispersion analytical component designed for precise measurements. It is suitable for use with TUV detectors and features specific dimensions for optimal performance.
  • Low-dispersion analytical flow cell
  • 10 mm path length
  • 500 nL illuminated volume
  • 63.5 μm PEEK interconnect inlet/outlet tubing
  • Designed for use with TUV detectors
Catalog No. 50-179-743 Supplier Waters Corp Supplier No. 205015033
Add to Cart
Add to Cart
Request a Quote
This TUV flow cell is a low-dispersion analytical component designed for precise measurements. It is suitable for use with TUV detectors and features specific dimensions for optimal performance.
  • Low-dispersion analytical flow cell
  • 10 mm path length
  • 500 nL illuminated volume
  • 63.5 μm PEEK interconnect inlet/outlet tubing
  • Designed for use with TUV detectors