Learn More
Waters Corp ACQUITY UPC2 BEH Column, 2.1 x 75 mm, 1.7 μm, 130Å
Catalog No. 50466315 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY UPC2 BEH Column is designed for high-performance chiral and achiral separations. It offers excellent peak shapes and a wide range of selectivities, making it suitable for various laboratory applications.
  • Designed for chiral and achiral separations.
  • Provides excellent peak shapes and unique selectivities.
  • Offers high efficiency and QC-ready robustness.
  • Features optimized particle size, column dimension, and flow rates.
  • Compatible with mass spectrometry detection.
  • Enables faster method development.
  • Ensures high quality, consistent, and reproducible results.
  • Uses inexpensive and non-toxic compressed liquid CO2 as a primary mobile phase, reducing solvent costs.
  • Designed for method robustness and extended column lifetimes.
  • Offers enhanced resolution for complex hydrophobic samples.
Catalog No. 50-466-315 Supplier Waters Corp Supplier No. 186006559
The ACQUITY UPC2 BEH Column is designed for high-performance chiral and achiral separations. It offers excellent peak shapes and a wide range of selectivities, making it suitable for various laboratory applications.
  • Designed for chiral and achiral separations.
  • Provides excellent peak shapes and unique selectivities.
  • Offers high efficiency and QC-ready robustness.
  • Features optimized particle size, column dimension, and flow rates.
  • Compatible with mass spectrometry detection.
  • Enables faster method development.
  • Ensures high quality, consistent, and reproducible results.
  • Uses inexpensive and non-toxic compressed liquid CO2 as a primary mobile phase, reducing solvent costs.
  • Designed for method robustness and extended column lifetimes.
  • Offers enhanced resolution for complex hydrophobic samples.