Learn More
Waters Corp Waters Column, 130Å, 1.7 μm, 3 mm X 100 mm
Catalog No. 50466320 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column is designed and tested for the ACQUITY UPC2 system, providing high efficiency and resolution. It uses sub-2-μm and 3.5 μm particles with controlled surface silanol activity for excellent peak shapes under SFC conditions without mobile-phase additives. Ethylene-Bridged Hybrid (BEH) and Charged-Surface Hybrid (CSH) particle technologies ensure broad selectivities and batch-to-batch repeatability.
  • Designed for ACQUITY UPC2 system for high efficiency and resolution
  • Controlled surface silanol activity for excellent peak shapes in SFC
  • Combines BEH and CSH particle technologies for broad selectivities and batch-to-batch repeatability
  • 1.7 μm particle size; 130 Å pore size
  • Maximum pressure: 6000 psi (415 bar)
  • 3 mm inner diameter; 100 mm length
  • Spherical particle shape
  • One unit per package
Catalog No. 50-466-320 Supplier Waters Corp Supplier No. 186006564
This column is designed and tested for the ACQUITY UPC2 system, providing high efficiency and resolution. It uses sub-2-μm and 3.5 μm particles with controlled surface silanol activity for excellent peak shapes under SFC conditions without mobile-phase additives. Ethylene-Bridged Hybrid (BEH) and Charged-Surface Hybrid (CSH) particle technologies ensure broad selectivities and batch-to-batch repeatability.
  • Designed for ACQUITY UPC2 system for high efficiency and resolution
  • Controlled surface silanol activity for excellent peak shapes in SFC
  • Combines BEH and CSH particle technologies for broad selectivities and batch-to-batch repeatability
  • 1.7 μm particle size; 130 Å pore size
  • Maximum pressure: 6000 psi (415 bar)
  • 3 mm inner diameter; 100 mm length
  • Spherical particle shape
  • One unit per package