Learn More
Waters Corp Waters BEH Column, 130Å, 1.7 μm, 2.1 mm x 50 mm
Catalog No. 50466314 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column is designed and tested for the ACQUITY UPC2 system, offering high efficiency and resolution. The controlled surface silanol activity on its particles ensures excellent peak shapes under SFC conditions — even for well-retained basic achiral compounds — without the need for mobile-phase additives.
  • Designed and tested for the ACQUITY UPC2 system.
  • Demonstrates high efficiency and resolution.
  • Reduction and control of surface silanol activity delivers excellent peak shapes under SFC conditions without the use of mobile-phase additives.
  • Effective even for well-retained basic achiral compounds.
Catalog No. 50-466-314 Supplier Waters Corp Supplier No. 186006558
This column is designed and tested for the ACQUITY UPC2 system, offering high efficiency and resolution. The controlled surface silanol activity on its particles ensures excellent peak shapes under SFC conditions — even for well-retained basic achiral compounds — without the need for mobile-phase additives.
  • Designed and tested for the ACQUITY UPC2 system.
  • Demonstrates high efficiency and resolution.
  • Reduction and control of surface silanol activity delivers excellent peak shapes under SFC conditions without the use of mobile-phase additives.
  • Effective even for well-retained basic achiral compounds.