Learn More
Waters Corp Viridis BEH 2-Ethylpyridine Column, 130Å, 1.7 μm
Catalog No. 50466415 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Viridis BEH 2-Ethylpyridine Column is designed for the ACQUITY UPC2 system, offering high efficiency and resolution for supercritical fluid chromatography (SFC) applications. It provides excellent peak shapes for various compounds without mobile-phase additives, ensuring high reproducibility and consistent performance due to controlled surface silanol activity and low metal content. These columns are suitable for preparative analytical and purification applications.
  • High efficiency and resolution for SFC separations.
  • Excellent peak shapes without mobile-phase additives.
  • Good retention, peak shape, and selectivity.
  • Exceptional batch-to-batch reproducibility.
  • Low metal content.
  • Chromatographically tested packing material with acidic, basic, and neutral analytes.
  • eCord™ Intelligent Chip with Performance Chromatogram and Certificate of Batch Analysis.
Catalog No. 50-466-415 Supplier Waters Corp Supplier No. 186006688
The Viridis BEH 2-Ethylpyridine Column is designed for the ACQUITY UPC2 system, offering high efficiency and resolution for supercritical fluid chromatography (SFC) applications. It provides excellent peak shapes for various compounds without mobile-phase additives, ensuring high reproducibility and consistent performance due to controlled surface silanol activity and low metal content. These columns are suitable for preparative analytical and purification applications.
  • High efficiency and resolution for SFC separations.
  • Excellent peak shapes without mobile-phase additives.
  • Good retention, peak shape, and selectivity.
  • Exceptional batch-to-batch reproducibility.
  • Low metal content.
  • Chromatographically tested packing material with acidic, basic, and neutral analytes.
  • eCord™ Intelligent Chip with Performance Chromatogram and Certificate of Batch Analysis.