Learn More
Waters Corp Viridis BEH 2-Ethylpyridine Column, 130Å, 3.5 μm, 2.1 mm X 150 mm
Catalog No. 50466406 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column is for the ACQUITY UPC2 system, offering high efficiency and resolution for SuperCritical Fluid Chromatography (SFC). It employs patented Ethylene Bridged Hybrid (BEH) particle technology for excellent peak shapes, especially for well-retained basic achiral compounds, often without mobile-phase additives.
  • Designed and tested for the ACQUITY UPC2 system.
  • Provides high efficiency and resolution for SFC separations.
  • Achieves excellent peak shapes via controlled surface silanol activity, reducing need for mobile-phase additives.
  • Effective for well-retained basic achiral compounds.
  • Enables fine-tuning of resolving power and selectivity for various mixtures.
  • Suitable for both chiral and achiral separations.
Catalog No. 50-466-406 Supplier Waters Corp Supplier No. 186006655
Add to Cart
Add to Cart
This column is for the ACQUITY UPC2 system, offering high efficiency and resolution for SuperCritical Fluid Chromatography (SFC). It employs patented Ethylene Bridged Hybrid (BEH) particle technology for excellent peak shapes, especially for well-retained basic achiral compounds, often without mobile-phase additives.
  • Designed and tested for the ACQUITY UPC2 system.
  • Provides high efficiency and resolution for SFC separations.
  • Achieves excellent peak shapes via controlled surface silanol activity, reducing need for mobile-phase additives.
  • Effective for well-retained basic achiral compounds.
  • Enables fine-tuning of resolving power and selectivity for various mixtures.
  • Suitable for both chiral and achiral separations.