Learn More
Waters Corp WATERS CORPORATION ACQUITY UPC2 BEH 2-EP Column, 1.7 μm, 2.1 x 50 mm
Catalog No. 50466330 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This ACQUITY UPC2 BEH 2-EP column is designed for a wide range of applications with acidic and basic compounds, providing specific characteristics for peak shape, retention, and selectivity.
  • Designed for a wide range of applications with acidic and basic compounds.
  • Provides specific characteristics for peak shape, retention, and selectivity.
  • Dimensions of 2.1 x 50 mm.
  • Particle size of 1.7 μm.
Catalog No. 50-466-330 Supplier Waters Corp Supplier No. 186006576
This ACQUITY UPC2 BEH 2-EP column is designed for a wide range of applications with acidic and basic compounds, providing specific characteristics for peak shape, retention, and selectivity.
  • Designed for a wide range of applications with acidic and basic compounds.
  • Provides specific characteristics for peak shape, retention, and selectivity.
  • Dimensions of 2.1 x 50 mm.
  • Particle size of 1.7 μm.