Learn More
Waters Corp ACQUITY UPC2 BEH 2-EP Column, 3.0 x 75 mm, 3.5 μm, BEH 2-Ethylpyridine
Catalog No. 50466408 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY UPC2 BEH 2-EP columns are designed for general use, suitable for a wide range of applications with acidic and basic compounds. This phase offers enhanced performance compared to conventional 2-ethylpyridine (2-EP), displaying improved peak shape, added retention, and novel selectivity. These columns provide flexibility for method development, usable with or without additives, maintaining exceptional peak shape.
  • Designed for general use
  • Suitable for acidic and basic compounds
  • Enhanced performance over conventional 2-ethylpyridine (2-EP)
  • Improved peak shape, added retention, and novel selectivity
  • Flexible for method development (with or without additives)
  • Maintains exceptional peak shape
Catalog No. 50-466-408 Supplier Waters Corp Supplier No. 186006657
ACQUITY UPC2 BEH 2-EP columns are designed for general use, suitable for a wide range of applications with acidic and basic compounds. This phase offers enhanced performance compared to conventional 2-ethylpyridine (2-EP), displaying improved peak shape, added retention, and novel selectivity. These columns provide flexibility for method development, usable with or without additives, maintaining exceptional peak shape.
  • Designed for general use
  • Suitable for acidic and basic compounds
  • Enhanced performance over conventional 2-ethylpyridine (2-EP)
  • Improved peak shape, added retention, and novel selectivity
  • Flexible for method development (with or without additives)
  • Maintains exceptional peak shape