Learn More
Waters Corp Viridis BEH 2-Ethylpyridine VanGuard Pre-column, 130Å, 3.5 µm, 2.1 mm X 5 mm, 3/pk
Catalog No. 50466402 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Designed and tested for the ACQUITY UPC2 system, the Viridis sub-2-µm and 3.5 µm columns demonstrate high efficiency and resolution. The reduction and control of surface silanol activity on Viridis particles delivers, under SFC conditions, excellent peak shapes — without the use of mobile-phase additives — even for well-retained basic achiral compounds.
Catalog No. 50-466-402 Supplier Waters Corp Supplier No. 186006651
Designed and tested for the ACQUITY UPC2 system, the Viridis sub-2-µm and 3.5 µm columns demonstrate high efficiency and resolution. The reduction and control of surface silanol activity on Viridis particles delivers, under SFC conditions, excellent peak shapes — without the use of mobile-phase additives — even for well-retained basic achiral compounds.