Learn More
Waters Corp Waters Column, 2-Ethylpyridine, 1.7 μm, 2.1 mm × 75 mm
Catalog No. 50466331 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Designed and tested for the ACQUITY UPC2 system, this column demonstrates high efficiency and resolution. The reduction and control of surface silanol activity on particles delivers, under SFC conditions, excellent peak shapes — without the use of mobile-phase additives — even for well-retained basic achiral compounds.
  • Designed and tested for the ACQUITY UPC2 system.
  • Columns demonstrate high efficiency and resolution.
  • Reduction and control of surface silanol activity on particles delivers excellent peak shapes under SFC conditions.
  • No mobile-phase additives required, even for well-retained basic achiral compounds.
Catalog No. 50-466-331 Supplier Waters Corp Supplier No. 186006577
Designed and tested for the ACQUITY UPC2 system, this column demonstrates high efficiency and resolution. The reduction and control of surface silanol activity on particles delivers, under SFC conditions, excellent peak shapes — without the use of mobile-phase additives — even for well-retained basic achiral compounds.
  • Designed and tested for the ACQUITY UPC2 system.
  • Columns demonstrate high efficiency and resolution.
  • Reduction and control of surface silanol activity on particles delivers excellent peak shapes under SFC conditions.
  • No mobile-phase additives required, even for well-retained basic achiral compounds.