Learn More
Waters Corp Waters ACQUITY UPC2 BEH Column, 130Å, 3.5 μm, 2.1 x 50 mm
Supplier:  Waters Corp 186006634
Encompass_Preferred
This ACQUITY UPC2 BEH column is designed for chiral and achiral separations and is compatible with the ACQUITY UPC2 System. It utilizes BEH particle chemistry to provide robust chromatographic performance.
  • Designed for chiral and achiral separations
  • Part of the ACQUITY UPC2 System
  • Enables precise variation of mobile phase strength, pressure, and temperature
  • Uses inexpensive and non-toxic compressed liquid CO2 as a primary mobile phase
  • Capable of separating challenging compounds, including structural analogs, isomers, and enantiomeric and diasteriomeric mixtures
  • Features BEH particle chemistry for robust chromatographic performance
Catalog No. 50-466-385