Learn More
Waters Corp Viridis Column, BEH 130Å, 3.5 μm, 2.1 x 100 mm
Catalog No. 50466387 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column is designed for use with a specific supercritical fluid chromatography (SFC) system, offering high efficiency and resolution. It features controlled surface silanol activity, which leads to excellent peak shapes in SFC conditions, even for certain challenging compounds, without requiring mobile-phase additives.
  • Designed and tested for high-performance SFC systems.
  • Delivers high efficiency and resolution.
  • Controlled surface silanol activity ensures excellent peak shapes.
  • Operates effectively without the need for mobile-phase additives for many compounds.
  • Suitable for well-retained basic achiral compounds under SFC conditions.
Catalog No. 50-466-387 Supplier Waters Corp Supplier No. 186006636
This column is designed for use with a specific supercritical fluid chromatography (SFC) system, offering high efficiency and resolution. It features controlled surface silanol activity, which leads to excellent peak shapes in SFC conditions, even for certain challenging compounds, without requiring mobile-phase additives.
  • Designed and tested for high-performance SFC systems.
  • Delivers high efficiency and resolution.
  • Controlled surface silanol activity ensures excellent peak shapes.
  • Operates effectively without the need for mobile-phase additives for many compounds.
  • Suitable for well-retained basic achiral compounds under SFC conditions.