Learn More
Waters Corp Column, BEH, 130Å Pore, 3.5 μm Particle, 3 mm ID, 100 mm Length
Supplier:  Waters Corp 186006640
Encompass_Preferred
These columns, designed for the ACQUITY UPC2 system, offer high efficiency and resolution for both chiral and achiral separations. They provide excellent peak shapes under SuperCritical Fluid (SFC) conditions without mobile-phase additives, achieving this by reducing and controlling surface silanol activity.
  • Enables precise variation of mobile phase strength, pressure, and temperature for challenging mixtures.
  • Utilizes inexpensive and non-toxic compressed liquid CO2 as a primary mobile phase.
  • Compatible with both polar and non-polar stationary phases.
  • Supports mass spectrometry detection in everyday laboratory use.
  • Reduces toxic solvent use and provides cost savings.
Catalog No. 50-466-391