Learn More
Waters Corp Performance Maintenance Kit, ACQUITY UPC2 Convergence Manager, Annual
Catalog No. 502392269 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Performance Maintenance Kit for the ACQUITY UPC² Convergence Manager contains all the quality parts necessary to keep the instrument operating at peak performance on an annual basis.
  • Includes an air chassis filter
  • Includes an inject valve pod
  • Includes filters
  • Includes a valve cartridge
  • Designed for annual maintenance
  • Ensures instrument operates at peak performance
Catalog No. 50-239-2269 Supplier Waters Corp Supplier No. 201000272
This Performance Maintenance Kit for the ACQUITY UPC² Convergence Manager contains all the quality parts necessary to keep the instrument operating at peak performance on an annual basis.
  • Includes an air chassis filter
  • Includes an inject valve pod
  • Includes filters
  • Includes a valve cartridge
  • Designed for annual maintenance
  • Ensures instrument operates at peak performance