Learn More
Waters Corp Waters Corporation Column, CSH Fluoro-Phenyl, 3.0 x 50 mm, 1.7 μm, 130 Å
Catalog No. 50466326 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This ACQUITY UPC2 CSH Fluoro-Phenyl Column is designed for efficient chiral and achiral separations, providing both speed and confidence in results. It integrates with the ACQUITY UPC2 System for convergence chromatography, utilizing inexpensive and non-toxic compressed liquid CO2 as a primary mobile phase. The column is compatible with mass spectrometry detection and offers a wide polarity range with a simple retention mechanism.
  • Handles chiral and achiral separations with speed and confidence.
  • Leverages chromatographic principles and selectivity of normal-phase chromatography.
  • Provides ease-of-use and method development simplicity similar to reversed-phase LC.
  • Uses inexpensive and non-toxic compressed liquid CO2 as a primary mobile phase.
  • Compatible with mass spectrometry detection.
Catalog No. 50-466-326 Supplier Waters Corp Supplier No. 186006571
This ACQUITY UPC2 CSH Fluoro-Phenyl Column is designed for efficient chiral and achiral separations, providing both speed and confidence in results. It integrates with the ACQUITY UPC2 System for convergence chromatography, utilizing inexpensive and non-toxic compressed liquid CO2 as a primary mobile phase. The column is compatible with mass spectrometry detection and offers a wide polarity range with a simple retention mechanism.
  • Handles chiral and achiral separations with speed and confidence.
  • Leverages chromatographic principles and selectivity of normal-phase chromatography.
  • Provides ease-of-use and method development simplicity similar to reversed-phase LC.
  • Uses inexpensive and non-toxic compressed liquid CO2 as a primary mobile phase.
  • Compatible with mass spectrometry detection.