Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters VanGuard Pre-Column, CSH Fluoro-Phenyl, 3.5 μm, 2.1 x 5 mm

Supplier:  Waters Corp 186006642

Encompass_Preferred

The VanGuard Pre-Column, CSH Fluoro-Phenyl, is designed for use with ACQUITY UPC2 systems. It serves to protect analytical columns by removing particulate matter and chemical contaminants, thereby extending column lifetime and improving system performance.

  • Dimensions: 2.1 x 5 mm.
  • Particle size: 3.5 μm.
  • Column type: CSH Fluoro-Phenyl.
  • Functions as a pre-column for system protection.

Catalog No. 50-466-393


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.