Learn More
Waters Corp Viridis Column, 130Å, 1.7 μm, 2.1 mm X 150 mm
Catalog No. 50466325 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column is designed and tested for the ACQUITY UPC2 system and features sub-2-μm and 3.5 μm particles, demonstrating high efficiency and resolution. The reduction and control of surface silanol activity on Viridis particles enable excellent peak shapes under SuperCritical Fluid (SFC) conditions, even for well-retained basic achiral compounds, without the need for mobile-phase additives.
  • Designed and tested for the ACQUITY UPC2 system.
  • Sub-2-μm and 3.5 μm columns demonstrate high efficiency and resolution.
  • Reduction and control of surface silanol activity on particles delivers excellent peak shapes under SFC conditions.
  • Achieves excellent peak shapes without mobile-phase additives, even for well-retained basic achiral compounds.
Catalog No. 50-466-325 Supplier Waters Corp Supplier No. 186006570
Add to Cart
Add to Cart
This column is designed and tested for the ACQUITY UPC2 system and features sub-2-μm and 3.5 μm particles, demonstrating high efficiency and resolution. The reduction and control of surface silanol activity on Viridis particles enable excellent peak shapes under SuperCritical Fluid (SFC) conditions, even for well-retained basic achiral compounds, without the need for mobile-phase additives.
  • Designed and tested for the ACQUITY UPC2 system.
  • Sub-2-μm and 3.5 μm columns demonstrate high efficiency and resolution.
  • Reduction and control of surface silanol activity on particles delivers excellent peak shapes under SFC conditions.
  • Achieves excellent peak shapes without mobile-phase additives, even for well-retained basic achiral compounds.