Learn More
Waters Corp Column, CSH Fluoro-Phenyl, 3.5 μm, 2.1 x 75 mm
Supplier:  Waters Corp 186006644
Encompass_Preferred
This column is part of the ACQUITY UPC2 column family, designed for advanced chromatographic separations. It provides precise control over separation parameters, enabling the analysis of challenging compounds.
  • Enables precise variation of mobile phase strength, pressure, and temperature.
  • Allows fine-tuning of resolving power and selectivity.
  • Facilitates better control over analyte retention for separating, detecting, and quantifying structural analogs, isomers, and enantiomeric and diastereomeric mixtures.
  • Utilizes inexpensive and non-toxic compressed liquid CO2 as a primary mobile phase.
  • Offers liquid CO2-based separations, providing performance and reliability.
  • Combines the ease-of-use of reversed-phase LC with the separation power of normal-phase LC through convergence chromatography.
Catalog No. 50-466-395