Learn More
Waters Corp Viridis CSH Fluoro-Phenyl Column, 130Å, 3.5 μm, 3 × 150 mm
Supplier:  Waters Corp 186006650
Encompass_Preferred
This CSH Fluoro-Phenyl Column is designed and tested for high-efficiency SuperCritical Fluid (SFC) separations. It features sub-2-μm and 3.5 μm columns that provide high efficiency and resolution. The product is engineered with reduced and controlled surface silanol activity, which helps deliver excellent peak shapes under SFC conditions, even without the need for mobile-phase additives. This characteristic is particularly beneficial for well-retained basic achiral compounds.
  • High efficiency and resolution
  • Excellent peak shapes under SFC conditions
  • Achieves excellent peak shapes without mobile-phase additives
  • Effective for well-retained basic achiral compounds
  • Reduction and control of surface silanol activity
Catalog No. 50-466-401