Learn More
Waters Corp Waters Analytical Column, Viridis CSH Fluoro-Phenyl, 1.7 μm, 3.0 × 150 mm
Catalog No. 50466414 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This analytical column is designed for achiral SFC separations. It utilizes CSH fluoro-phenyl chemistry to provide excellent performance in various SFC applications.
  • Particle size of 1.7 μm
  • Features CSH fluoro-phenyl chemistry with 10% carbon load
  • Suitable for SFC separations
  • Provides good retention, peak shape, and selectivity in achiral SFC separations
  • Can be used both with and without additives
  • Part of the Viridis analytical and preparative achiral SFC columns line
Catalog No. 50-466-414 Supplier Waters Corp Supplier No. 186006687
This analytical column is designed for achiral SFC separations. It utilizes CSH fluoro-phenyl chemistry to provide excellent performance in various SFC applications.
  • Particle size of 1.7 μm
  • Features CSH fluoro-phenyl chemistry with 10% carbon load
  • Suitable for SFC separations
  • Provides good retention, peak shape, and selectivity in achiral SFC separations
  • Can be used both with and without additives
  • Part of the Viridis analytical and preparative achiral SFC columns line