Learn More
Waters Corp Waters Corporation Viridis CSH Fluoro-Phenyl, 3.0 x 75 mm, 3.5 μm
Catalog No. 50466399 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This product is a Viridis CSH Fluoro-Phenyl, designed for advanced chromatographic separations. It features CSH particle technology, ensuring excellent peak shapes and consistent performance.
  • Based on Charged Surface Hybrid (CSH) particle technology.
  • Offers an added range of achiral SFC selectivities.
  • Reduction and control of surface silanol activity.
  • Delivers excellent peak shapes even for well-retained basic achiral compounds under SFC conditions.
  • Dimensions: 3.0 x 75 mm.
  • Particle size: 3.5 μm.
Catalog No. 50-466-399 Supplier Waters Corp Supplier No. 186006648
This product is a Viridis CSH Fluoro-Phenyl, designed for advanced chromatographic separations. It features CSH particle technology, ensuring excellent peak shapes and consistent performance.
  • Based on Charged Surface Hybrid (CSH) particle technology.
  • Offers an added range of achiral SFC selectivities.
  • Reduction and control of surface silanol activity.
  • Delivers excellent peak shapes even for well-retained basic achiral compounds under SFC conditions.
  • Dimensions: 3.0 x 75 mm.
  • Particle size: 3.5 μm.