Learn More
Waters Corp Waters Corporation Column, PFP/Fluoro-Phenyl, 1.7 μm, 3 mm X 75 mm, 130 Å
Catalog No. 50466327 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Designed and tested for the ACQUITY UPC2 system, these columns demonstrate high efficiency and resolution. The reduction and control of surface silanol activity on the particles delivers excellent peak shapes under SFC conditions, even for well-retained basic achiral compounds, without the use of mobile-phase additives.
  • Designed and tested for the ACQUITY UPC2 system
  • Demonstrates high efficiency and resolution
  • Reduction and control of surface silanol activity on particles
  • Delivers excellent peak shapes under SFC conditions
  • Eliminates the need for mobile-phase additives
  • Suitable for well-retained basic achiral compounds
Catalog No. 50-466-327 Supplier Waters Corp Supplier No. 186006572
Add to Cart
Add to Cart
Designed and tested for the ACQUITY UPC2 system, these columns demonstrate high efficiency and resolution. The reduction and control of surface silanol activity on the particles delivers excellent peak shapes under SFC conditions, even for well-retained basic achiral compounds, without the use of mobile-phase additives.
  • Designed and tested for the ACQUITY UPC2 system
  • Demonstrates high efficiency and resolution
  • Reduction and control of surface silanol activity on particles
  • Delivers excellent peak shapes under SFC conditions
  • Eliminates the need for mobile-phase additives
  • Suitable for well-retained basic achiral compounds