Learn More
Waters Corp ACQUITY UPC2 HSS C18 SB Column, 3.5 μm, 100 Å, 3 x 50 mm
Catalog No. 50466380 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters ACQUITY UPC2 HSS C18 SB columns utilize Convergence Chromatography (UPC2) with compressed CO2 as the primary mobile phase. This technique combines the selectivity of normal-phase chromatography with the ease of use of reversed-phase LC, allowing for separations across a wide polarity range, particularly effective for challenging separations of structural analogs, isomers, and mixtures.
  • Uses inexpensive and non-toxic compressed liquid CO2 as a primary mobile phase, reducing toxic solvent use and associated costs.
  • Offers precise variation of mobile phase strength, pressure, and temperature for fine-tuning separation selectivity.
  • Provides full mass spectrometry detection capabilities, which was previously challenging with normal-phase LC.
  • Enables faster method development protocols.
  • Delivers high quality, consistent, and reproducible columns.
Catalog No. 50-466-380 Supplier Waters Corp Supplier No. 186006629
Waters ACQUITY UPC2 HSS C18 SB columns utilize Convergence Chromatography (UPC2) with compressed CO2 as the primary mobile phase. This technique combines the selectivity of normal-phase chromatography with the ease of use of reversed-phase LC, allowing for separations across a wide polarity range, particularly effective for challenging separations of structural analogs, isomers, and mixtures.
  • Uses inexpensive and non-toxic compressed liquid CO2 as a primary mobile phase, reducing toxic solvent use and associated costs.
  • Offers precise variation of mobile phase strength, pressure, and temperature for fine-tuning separation selectivity.
  • Provides full mass spectrometry detection capabilities, which was previously challenging with normal-phase LC.
  • Enables faster method development protocols.
  • Delivers high quality, consistent, and reproducible columns.