Learn More
Waters Corp Viridis HSS C18 SB Column, 100Å, 3.5 µm, 2.1 mm X 50 mm, 1/pk
Catalog No. 50466376 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Viridis sub-2-µm and 3.5 µm columns demonstrate high efficiency and resolution. The reduction and control of surface silanol activity on Viridis particles delivers, under SFC conditions, excellent peak shapes — without the use of mobile-phase additives — even for well-retained basic achiral compounds.
Catalog No. 50-466-376 Supplier Waters Corp Supplier No. 186006625
The Viridis sub-2-µm and 3.5 µm columns demonstrate high efficiency and resolution. The reduction and control of surface silanol activity on Viridis particles delivers, under SFC conditions, excellent peak shapes — without the use of mobile-phase additives — even for well-retained basic achiral compounds.