Learn More
Waters Corp ACQUITY UPC2 HSS C18 SB Column, 100Å, 3.5 μm, 3 mm x 75 mm
Catalog No. 50466381 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY UPC2 HSS C18 SB Column is designed for use with the ACQUITY UPC2 System, leveraging compressed liquid CO2 for efficient, cost-effective separations. It offers precise mobile phase control for enhanced resolving power across diverse compounds.
  • Precisely vary mobile phase strength, pressure, and temperature.
  • Separates structural analogs, isomers, and enantiomeric/diasteriomeric mixtures.
  • Uses inexpensive and non-toxic compressed liquid CO2 as a primary mobile phase.
  • Combines reversed-phase LC ease-of-use with normal-phase LC separation power.
  • Mass spectrometry compatible.
  • Reduces toxic solvent use and offers cost savings.
  • Optimized particle size, column dimension, and flow rates ensure high quality and consistent performance.
  • Designed for excellent peak shapes without additives and extended column lifetimes.
Catalog No. 50-466-381 Supplier Waters Corp Supplier No. 186006630
The ACQUITY UPC2 HSS C18 SB Column is designed for use with the ACQUITY UPC2 System, leveraging compressed liquid CO2 for efficient, cost-effective separations. It offers precise mobile phase control for enhanced resolving power across diverse compounds.
  • Precisely vary mobile phase strength, pressure, and temperature.
  • Separates structural analogs, isomers, and enantiomeric/diasteriomeric mixtures.
  • Uses inexpensive and non-toxic compressed liquid CO2 as a primary mobile phase.
  • Combines reversed-phase LC ease-of-use with normal-phase LC separation power.
  • Mass spectrometry compatible.
  • Reduces toxic solvent use and offers cost savings.
  • Optimized particle size, column dimension, and flow rates ensure high quality and consistent performance.
  • Designed for excellent peak shapes without additives and extended column lifetimes.