Learn More
Waters Corp Waters Column, C18, 100Å, 1.8 μm, 2.1 mm x 100 mm
Supplier:  Waters Corp 186006619
Encompass_Preferred
This column is designed and tested for the ACQUITY UPC2 system, demonstrating high efficiency and resolution. The reduction and control of surface silanol activity on Viridis particles delivers excellent peak shapes under SFC conditions, even for well-retained basic achiral compounds, without the use of mobile-phase additives.
  • Designed and tested for the ACQUITY UPC2 system.
  • Demonstrates high efficiency and resolution.
  • Reduction and control of surface silanol activity on HSS particles delivers excellent peak shapes under SFC conditions.
  • Achieves excellent peak shapes without the use of mobile-phase additives, even for well-retained basic achiral compounds.
Catalog No. 50-466-370