Learn More
Waters Corp Viridis Column, HSS C18 SB, 3.5 µm, 2.1 mm, 75 mm
Supplier:  Waters Corp 186006626
Encompass_Preferred
  • Designed and tested for the ACQUITY UPC2 system, offering high efficiency and resolution.
  • Features reduced and controlled surface silanol activity, delivering excellent peak shapes without mobile-phase additives.
  • Provides mass spectrometry compatibility using compressed liquid CO2 as the primary mobile phase.
  • Reduces toxic solvent use and associated costs.
  • Enables separation of a broad spectrum of compounds across a wide polarity range.
  • Ensures robust and consistent performance with reproducible results and stable selectivity.
  • Enhances separation of challenging compounds, including structural analogs, isomers, and mixtures.
  • Facilitates faster method development through optimized parameters.
  • Offers cost-effectiveness due to reduced solvent consumption and faster analysis times.
Catalog No. 50-466-377